Product Details

SNP ID
rs17478818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:24998631 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATTCCACAGATCAGGCAGAGTGT[A/G]ACCATGAAGCTCTTCAGTGAATTTT
Phenotype
MIM: 608301
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LGI2 PubMed Links

Gene Details

Gene
LGI2
Gene Name
leucine rich repeat LGI family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018176.3 Intron NP_060646.2
XM_011513850.2 Intron XP_011512152.1
XM_017008356.1 Intron XP_016863845.1

View Full Product Details