Product Details
- SNP ID
-
rs11849590
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:75578902 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCCTTTCAACTCTAAGAGACCAG[C/T]AGAGGCCACTGTCCCTTAAGACTGC
- Phenotype
-
MIM: 610865
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FLVCR2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148092740] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FLVCR2
- Gene Name
- feline leukemia virus subgroup C cellular receptor family member 2
- Gene
- LOC102724153
- Gene Name
- uncharacterized LOC102724153
There are no transcripts associated with this gene.
View Full Product Details