Product Details

SNP ID
rs3818572
Assay Type
Validated
NCBI dbSNP Submissions
39
Location
Chr.1:108650634 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
AAGGAAAATTATGGGCAGGGTGTAG[C/G]GGTGGACAAGGAGGCAACAAGTGCT
Phenotype
MIM: 612178
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HENMT1 PubMed Links

Gene Details

Gene
HENMT1
Gene Name
HEN1 methyltransferase homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102592.1 Intron NP_001096062.1
NM_144584.2 Intron NP_653185.2
XM_005270411.1 Intron XP_005270468.1
XM_017000179.1 Intron XP_016855668.1

View Full Product Details