Product Details
- SNP ID
-
rs987359315
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:131862332 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GCGGTGGATGATGGGGTCTGCTGGC[A/G]TGAGAGCATGCGGCCGTGTGAGCAT
- Phenotype
-
MIM: 605044
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MED27
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs913509] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MED27
- Gene Name
- mediator complex subunit 27
View Full Product Details