Product Details

SNP ID
rs13300883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:126819269 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGCATACCTCATTTTGTGCTTCA[C/T]GGATATTGCATTTTTTTTTTTTTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZBTB43 PubMed Links

Gene Details

Gene
ZBTB43
Gene Name
zinc finger and BTB domain containing 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135776.1 Intron NP_001129248.1
NM_014007.3 Intron NP_054726.1
XM_005251835.3 Intron XP_005251892.1
XM_011518409.1 Intron XP_011516711.1
XM_011518411.2 Intron XP_011516713.1

View Full Product Details