Product Details

SNP ID
rs35501618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:68488296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATCCAAACATGTAATTATTTAT[A/G]ATATGCTTAAAGTGCACACACATCT
Phenotype
MIM: 139080
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC25A16 PubMed Links

Gene Details

Gene
SLC25A16
Gene Name
solute carrier family 25 member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324312.1 Intron NP_001311241.1
NM_001324313.1 Intron NP_001311242.1
NM_001324314.1 Intron NP_001311243.1
NM_001324315.1 Intron NP_001311244.1
NM_001324317.1 Intron NP_001311246.1
NM_152707.3 Intron NP_689920.1
XM_011540211.2 Intron XP_011538513.1
XM_017016695.1 Intron XP_016872184.1

View Full Product Details