Product Details

SNP ID
rs7855063
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:78301699 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ATCACAGAGAGCATCACGGAGTGAT[A/G]AATTAATAAATAACTTGCTCAGTGA
Phenotype
MIM: 610936
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSAT1 PubMed Links

Gene Details

Gene
PSAT1
Gene Name
phosphoserine aminotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021154.4 Intron NP_066977.1
NM_058179.3 Intron NP_478059.1

View Full Product Details