Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001083946.1 | 3577 | Intron | NP_001077415.1 | ||
NM_144736.4 | 3577 | Intron | NP_653337.1 | ||
XM_005264410.1 | 3577 | Intron | XP_005264467.1 | ||
XM_005264411.1 | 3577 | Intron | XP_005264468.1 | ||
XM_011532956.1 | 3577 | Intron | XP_011531258.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_005813.4 | 3577 | UTR 3 | NP_005804.1 | ||
XM_005264237.3 | 3577 | UTR 3 | XP_005264294.1 | ||
XM_005264238.3 | 3577 | UTR 3 | XP_005264295.1 | ||
XM_017003741.1 | 3577 | UTR 3 | XP_016859230.1 | ||
XM_017003742.1 | 3577 | UTR 3 | XP_016859231.1 | ||
XM_017003743.1 | 3577 | UTR 3 | XP_016859232.1 | ||
XM_017003744.1 | 3577 | UTR 3 | XP_016859233.1 |