Product Details

SNP ID
rs11941723
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:17810905 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCCGCTCTCTTCTCCCAAAGCGCTT[A/G]CGGCAGTCTCGCGGGATTTCCCCCT
Phenotype
MIM: 606280
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCAF16 PubMed Links

Gene Details

Gene
DCAF16
Gene Name
DDB1 and CUL4 associated factor 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017741.3 93 Intron NP_060211.3
XM_005248169.3 93 Intron XP_005248226.1
XM_005248171.3 93 Intron XP_005248228.1
XM_006713966.3 93 Intron XP_006714029.1
XM_017008327.1 93 Intron XP_016863816.1
XM_017008328.1 93 Intron XP_016863817.1
Gene
NCAPG
Gene Name
non-SMC condensin I complex subunit G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022346.4 93 UTR 5 NP_071741.2
XM_011513876.2 93 Intron XP_011512178.1
XM_017008543.1 93 Intron XP_016864032.1
XM_017008544.1 93 Intron XP_016864033.1

View Full Product Details