Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_020366.3 | 287 | Missense Mutation | CAG,CCG | Q96P | NP_065099.3 |
XM_005267879.2 | 287 | Intron | XP_005267936.1 | ||
XM_005267880.2 | 287 | Intron | XP_005267937.1 | ||
XM_005267881.3 | 287 | Intron | XP_005267938.1 | ||
XM_011536978.1 | 287 | Intron | XP_011535280.1 | ||
XM_011536979.1 | 287 | Intron | XP_011535281.1 | ||
XM_011536980.1 | 287 | Intron | XP_011535282.1 | ||
XM_011536981.1 | 287 | Intron | XP_011535283.1 | ||
XM_011536982.1 | 287 | Intron | XP_011535284.1 | ||
XM_017021473.1 | 287 | Intron | XP_016876962.1 |