Product Details

SNP ID
rs2863807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70829248 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTGGTGAGGGAGGGATCCCCTGC[C/T]CTAAATAGGACAAGATGGCCAAAAC
Phenotype
MIM: 604862
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD207 PubMed Links

Gene Details

Gene
CD207
Gene Name
CD207 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015717.4 Intron NP_056532.4
XM_011532874.1 Intron XP_011531176.1
XM_011532875.2 Intron XP_011531177.1
XM_011532876.2 Intron XP_011531178.1
Gene
CLEC4F
Gene Name
C-type lectin domain family 4 member F
There are no transcripts associated with this gene.

View Full Product Details