Product Details
- SNP ID
-
rs9657915
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:105655970 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGAAATGGGACCCTTTTAAGTTGC[A/G]GTGAGGGTTCTATTATGAAGACTAT
- Phenotype
-
MIM: 604491
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CBLB
PubMed Links
Gene Details
- Gene
- CBLB
- Gene Name
- Cbl proto-oncogene B
View Full Product Details