Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014714.3 | Intron | NP_055529.2 | |||
XM_005255725.4 | Intron | XP_005255782.1 | |||
XM_005255726.3 | Intron | XP_005255783.1 | |||
XM_006720990.3 | Intron | XP_006721053.1 | |||
XM_006720991.3 | Intron | XP_006721054.1 | |||
XM_006720992.3 | Intron | XP_006721055.1 | |||
XM_011522766.2 | Intron | XP_011521068.1 | |||
XM_011522767.2 | Intron | XP_011521069.1 | |||
XM_011522769.2 | Intron | XP_011521071.1 | |||
XM_011522771.2 | Intron | XP_011521073.1 | |||
XM_011522772.2 | Intron | XP_011521074.1 | |||
XM_017023910.1 | Intron | XP_016879399.1 | |||
XM_017023911.1 | Intron | XP_016879400.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_016111.3 | Intron | NP_057195.2 | |||
XM_011522773.2 | Intron | XP_011521075.1 | |||
XM_011522774.2 | Intron | XP_011521076.1 | |||
XM_011522775.2 | Intron | XP_011521077.1 | |||
XM_011522776.2 | Intron | XP_011521078.1 | |||
XM_011522777.2 | Intron | XP_011521079.1 | |||
XM_011522778.2 | Intron | XP_011521080.1 | |||
XM_017023914.1 | Intron | XP_016879403.1 |