Product Details

SNP ID
rs34536593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45898774 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCATCTCTACTTGAATTTCACCT[C/T]CTCAGAGAAGCCCTCCCTGATTACC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYPOP PubMed Links

Gene Details

Gene
MYPOP
Gene Name
Myb related transcription factor, partner of profilin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012643.3 Intron NP_001012661.1
XM_017026742.1 Intron XP_016882231.1
XM_017026743.1 Intron XP_016882232.1

View Full Product Details