Product Details

SNP ID
rs10403030
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45892720 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACCATCCACATACAGAAGCTATC[A/G]TCTCTGCCTTCAAATTAAATACATA
Phenotype
MIM: 615331
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRF2BP1 PubMed Links

Gene Details

Gene
IRF2BP1
Gene Name
interferon regulatory factor 2 binding protein 1
There are no transcripts associated with this gene.

Gene
MYPOP
Gene Name
Myb related transcription factor, partner of profilin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012643.3 Intron NP_001012661.1
XM_017026742.1 Intron XP_016882231.1
XM_017026743.1 Intron XP_016882232.1

View Full Product Details