Product Details

SNP ID
rs8107967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7907730 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGCCTGGTGGGCTCTGTCAGGGA[A/G]GGGGCCCACAGGACTCCTCGAGGGC
Phenotype
MIM: 612891 MIM: 603014
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC8E PubMed Links

Gene Details

Gene
LRRC8E
Gene Name
leucine rich repeat containing 8 family member E
There are no transcripts associated with this gene.

Gene
MAP2K7
Gene Name
mitogen-activated protein kinase kinase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297555.1 Intron NP_001284484.1
NM_001297556.1 Intron NP_001284485.1
NM_145185.3 Intron NP_660186.1
XM_006722800.2 Intron XP_006722863.1
Gene
TGFBR3L
Gene Name
transforming growth factor beta receptor 3 like
There are no transcripts associated with this gene.

View Full Product Details