Product Details

SNP ID
rs12610401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7890177 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCTGTGGGTGTGGAGGGTCTCA[C/G]TCTCCCACCCTTCCCCCAGCAGGAA
Phenotype
MIM: 612891
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LRRC8E PubMed Links

Gene Details

Gene
LRRC8E
Gene Name
leucine rich repeat containing 8 family member E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001268284.2 Intron NP_001255213.1
NM_001268285.2 Intron NP_001255214.1
NM_025061.5 Intron NP_079337.2
XM_011528319.2 Intron XP_011526621.1
Gene
LYPLA2P2
Gene Name
lysophospholipase II pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details