Product Details

SNP ID
rs11737943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:31643129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGGTGTGTTTGGAAGGCCCTCC[A/G]TGACCGCATCTGCTGTATAGGGGGT
Phenotype
MIM: 610697
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDZD2 PubMed Links

Gene Details

Gene
PDZD2
Gene Name
PDZ domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178140.3 Intron NP_835260.2
XM_005248269.4 Intron XP_005248326.1
XM_005248271.1 Intron XP_005248328.1
XM_005248272.3 Intron XP_005248329.1
XM_006714460.2 Intron XP_006714523.1
XM_011513992.2 Intron XP_011512294.1
XM_011513993.2 Intron XP_011512295.1
XM_011513994.2 Intron XP_011512296.1
XM_011513995.2 Intron XP_011512297.1
XM_011513996.2 Intron XP_011512298.1
XM_017009245.1 Intron XP_016864734.1
XM_017009246.1 Intron XP_016864735.1

View Full Product Details