Product Details
- SNP ID
-
rs11160531
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:99606442 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCACAGAGTTGCTCCCTGGAGATC[C/G]AAGCGTAAACGTTGGATACACCATA
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC85C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs151204955] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC85C
- Gene Name
- coiled-coil domain containing 85C
There are no transcripts associated with this gene.
- Gene
- HHIPL1
- Gene Name
- HHIP like 1
View Full Product Details