Product Details

SNP ID
rs34684319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110129102 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGTACCAAGTGAGTTTCTTCAGG[A/G]TGAAACTGTGGCTGACACCAATAAA
Phenotype
MIM: 605489
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFT81 PubMed Links

Gene Details

Gene
IFT81
Gene Name
intraflagellar transport 81
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143779.1 641 Missense Mutation GAT,GGT D134G NP_001137251.1
NM_014055.3 641 Missense Mutation GAT,GGT D134G NP_054774.2
NM_031473.3 641 Missense Mutation GAT,GGT D134G NP_113661.2
XM_011538232.2 641 Missense Mutation GAT,GGT D134G XP_011536534.1
XM_017019217.1 641 Missense Mutation GAT,GGT D134G XP_016874706.1
XM_017019218.1 641 UTR 5 XP_016874707.1

View Full Product Details