Product Details

SNP ID
rs743725
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:86889 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAGGAGCCTGGAAGGGATGGGTGGG[C/T]GTGAGCCCAACCTGACACCAGCCCC
Phenotype
MIM: 156565 MIM: 600928
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MPG PubMed Links

Gene Details

Gene
MPG
Gene Name
N-methylpurine DNA glycosylase
There are no transcripts associated with this gene.

Gene
NPRL3
Gene Name
NPR3 like, GATOR1 complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039476.2 Intron NP_001034565.1
NM_001077350.2 Intron NP_001070818.1
NM_001243247.1 Intron NP_001230176.1
NM_001243248.1 Intron NP_001230177.1
NM_001243249.1 Intron NP_001230178.1

View Full Product Details