Product Details

SNP ID
rs11749169
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:170359520 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTCGCTCACTGTCCTGGCAGTGAG[A/G]GGCTGAGCAAGCACTCTGGTCATGT
Phenotype
MIM: 604660
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNIP1 PubMed Links

Gene Details

Gene
KCNIP1
Gene Name
potassium voltage-gated channel interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034837.2 Intron NP_001030009.1
NM_001034838.2 Intron NP_001030010.1
NM_001278339.1 Intron NP_001265268.1
NM_001278340.1 Intron NP_001265269.1
NM_014592.3 Intron NP_055407.1
XM_017009407.1 Intron XP_016864896.1
XM_017009408.1 Intron XP_016864897.1

View Full Product Details