Product Details

SNP ID
rs1396149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78132581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGGGGGGGCTGGCCCAGGGCCCA[C/G]AGCGTGGCGACAACGCTGGGCGTGG
Phenotype
MIM: 605828 MIM: 605829
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMC6 PubMed Links

Gene Details

Gene
TMC6
Gene Name
transmembrane channel like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127198.2 Intron NP_001120670.1
NM_001321185.1 Intron NP_001308114.1
NM_007267.7 Intron NP_009198.4
XM_005256995.1 Intron XP_005257052.1
XM_011524255.1 Intron XP_011522557.1
XM_011524256.1 Intron XP_011522558.1
XM_011524257.2 Intron XP_011522559.1
XM_011524258.1 Intron XP_011522560.1
XM_017024107.1 Intron XP_016879596.1
XM_017024108.1 Intron XP_016879597.1
XM_017024109.1 Intron XP_016879598.1
Gene
TMC8
Gene Name
transmembrane channel like 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152468.4 Intron NP_689681.2
XM_011524402.2 Intron XP_011522704.1
XM_011524403.2 Intron XP_011522705.1
XM_011524404.2 Intron XP_011522706.1
XM_011524406.2 Intron XP_011522708.1
XM_011524409.2 Intron XP_011522711.1
XM_011524410.2 Intron XP_011522712.1
XM_011524411.2 Intron XP_011522713.1
XM_017024238.1 Intron XP_016879727.1
XM_017024239.1 Intron XP_016879728.1
XM_017024240.1 Intron XP_016879729.1
XM_017024241.1 Intron XP_016879730.1
XM_017024242.1 Intron XP_016879731.1
XM_017024243.1 Intron XP_016879732.1
XM_017024244.1 Intron XP_016879733.1

View Full Product Details