Product Details
- SNP ID
-
rs10490794
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:42711380 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCACAGTAAATTACTTGGTATCAG[A/C]AGACCTGGCTTTCGCCCTCTGCTGA
- Phenotype
-
MIM: 608116
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC13
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144146748] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC13
- Gene Name
- coiled-coil domain containing 13
- Gene
- HHATL
- Gene Name
- hedgehog acyltransferase-like
There are no transcripts associated with this gene.
View Full Product Details