Product Details
- SNP ID
-
rs10770822
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:21529305 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGTTTGAAAACAGGAACTTCAGAG[C/T]CCGTTAATAAAGGTTGAAATCATGA
- Phenotype
-
MIM: 138571
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GYS2
PubMed Links
Gene Details
- Gene
- GYS2
- Gene Name
- glycogen synthase 2
There are no transcripts associated with this gene.
- Gene
- SPX
- Gene Name
- spexin hormone
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_030572.3 |
|
Intron |
|
|
NP_085049.1 |
View Full Product Details