Product Details

SNP ID
rs258865
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:173611156 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTTTCCTGGAGATTCTTAAACT[G/T]TTGAGGAAAACATAAAATCTTGTCA
Phenotype
MIM: 616745
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BOD1 PubMed Links

Gene Details

Gene
BOD1
Gene Name
biorientation of chromosomes in cell division 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159651.1 Intron NP_001153123.1
NM_138369.2 Intron NP_612378.1
XM_017010017.1 Intron XP_016865506.1

View Full Product Details