Product Details

SNP ID
rs2291297
Assay Type
Validated
NCBI dbSNP Submissions
54
Location
Chr.1:37806988 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
CGAATGCCTACAAGCTGTAAGGCAA[A/G]GGGAAAGGGATCATGAGAAAGGTTG
Phenotype
MIM: 600172 MIM: 612276
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf122 PubMed Links

Gene Details

Gene
C1orf122
Gene Name
chromosome 1 open reading frame 122
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142726.1 Intron NP_001136198.1
NM_198446.2 Intron NP_940848.2
Gene
MANEAL
Gene Name
mannosidase endo-alpha like
There are no transcripts associated with this gene.

Gene
MTF1
Gene Name
metal regulatory transcription factor 1
There are no transcripts associated with this gene.

Gene
YRDC
Gene Name
yrdC N6-threonylcarbamoyltransferase domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024640.3 Intron NP_078916.3

View Full Product Details