Product Details

SNP ID
rs209238
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:130215321 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AATGACATTCAATGCACTTGTGAAT[T/G]CCCCGACGACACCTTATGGAGACGG
Phenotype
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF280C PubMed Links

Gene Details

Gene
ZNF280C
Gene Name
zinc finger protein 280C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017666.4 2054 Silent Mutation GGA,GGC G617G NP_060136.1
XM_006724765.3 2054 Silent Mutation GGA,GGC G568G XP_006724828.1

View Full Product Details