Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001011722.2 | 31 | Intron | NP_001011722.2 | ||
NM_001319837.1 | 31 | Intron | NP_001306766.1 | ||
NM_001319838.1 | 31 | Intron | NP_001306767.1 | ||
NM_001328124.1 | 31 | Intron | NP_001315053.1 | ||
NM_018125.3 | 31 | Intron | NP_060595.3 | ||
XM_005245923.2 | 31 | Intron | XP_005245980.2 | ||
XM_005245925.2 | 31 | Intron | XP_005245982.2 | ||
XM_005245927.2 | 31 | Intron | XP_005245984.2 | ||
XM_005245929.2 | 31 | Intron | XP_005245986.2 | ||
XM_006710728.1 | 31 | UTR 5 | XP_006710791.1 | ||
XM_006710729.1 | 31 | Intron | XP_006710792.1 | ||
XM_006710731.1 | 31 | UTR 5 | XP_006710794.1 | ||
XM_011541691.2 | 31 | Intron | XP_011539993.1 | ||
XM_011541692.2 | 31 | Intron | XP_011539994.1 | ||
XM_011541693.2 | 31 | Intron | XP_011539995.1 | ||
XM_011541695.2 | 31 | Intron | XP_011539997.1 | ||
XM_011541696.2 | 31 | Intron | XP_011539998.1 | ||
XM_017001617.1 | 31 | Intron | XP_016857106.1 | ||
XM_017001618.1 | 31 | Intron | XP_016857107.1 | ||
XM_017001619.1 | 31 | Intron | XP_016857108.1 | ||
XM_017001620.1 | 31 | Intron | XP_016857109.1 | ||
XM_017001621.1 | 31 | Intron | XP_016857110.1 | ||
XM_017001622.1 | 31 | Intron | XP_016857111.1 |