Product Details
- SNP ID
-
rs3114130
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:2236574 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCACATCCAAGGTGAGTCTCTCGG[C/T]TGCCCTGAGCTGGGGCTGGATGGGC
- Phenotype
-
MIM: 602622
MIM: 603022
MIM: 600305
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DNASE1L2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs186907134] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DNASE1L2
- Gene Name
- deoxyribonuclease I-like 2
- Gene
- E4F1
- Gene Name
- E4F transcription factor 1
- Gene
- ECI1
- Gene Name
- enoyl-CoA delta isomerase 1
There are no transcripts associated with this gene.
View Full Product Details