Product Details
- SNP ID
-
rs35968577
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:87880700 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATAATCTCGGTGAAAACATCTTAAT[A/G]TCCCATTTATCTGCAGAGTATGCTT
- Phenotype
-
MIM: 604281
MIM: 610821
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DBF4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77012126] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DBF4
- Gene Name
- DBF4 zinc finger
- Gene
- SLC25A40
- Gene Name
- solute carrier family 25 member 40
There are no transcripts associated with this gene.
View Full Product Details