Product Details

SNP ID
rs6010637
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63745109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCAGCTCCCTGGTAGCCATGCT[C/G]TCCCCCTGGGGGACTCGTGCCAGAG
Phenotype
MIM: 609809 MIM: 609493 MIM: 614639
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
LIME1 PubMed Links

Gene Details

Gene
LIME1
Gene Name
Lck interacting transmembrane adaptor 1
There are no transcripts associated with this gene.

Gene
SLC2A4RG
Gene Name
SLC2A4 regulator
There are no transcripts associated with this gene.

Gene
ZBTB46
Gene Name
zinc finger and BTB domain containing 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025224.3 3872 UTR 3 NP_079500.2
XM_005260195.4 3872 UTR 3 XP_005260252.1
XM_005260196.3 3872 UTR 3 XP_005260253.1
XM_005260197.4 3872 UTR 3 XP_005260254.1
XM_005260198.4 3872 UTR 3 XP_005260255.1
XM_006723700.3 3872 UTR 3 XP_006723763.1
XM_011528548.2 3872 UTR 3 XP_011526850.1
XM_011528549.2 3872 Intron XP_011526851.1
XM_017027667.1 3872 Intron XP_016883156.1
Gene
ZGPAT
Gene Name
zinc finger CCCH-type and G-patch domain containing
There are no transcripts associated with this gene.

View Full Product Details