Product Details

SNP ID
rs2509050
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119107201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGATGGGGCGAGGTGATCAAAG[A/G]TCCCGCCTATTTTCAACCCCGGAAC
Phenotype
MIM: 191350
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2CD2L PubMed Links

Gene Details

Gene
C2CD2L
Gene Name
C2CD2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290474.1 141 Intron NP_001277403.1
NM_014807.4 141 Intron NP_055622.3
XM_006718948.3 141 UTR 5 XP_006719011.1
XM_011543076.2 141 Intron XP_011541378.1
XM_017018625.1 141 UTR 5 XP_016874114.1
XM_017018626.1 141 UTR 5 XP_016874115.1
XM_017018627.1 141 UTR 5 XP_016874116.1
XM_017018628.1 141 Intron XP_016874117.1
Gene
DPAGT1
Gene Name
dolichyl-phosphate N-acetylglucosaminephosphotransferase 1
There are no transcripts associated with this gene.

View Full Product Details