Product Details

SNP ID
rs510435
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:119109329 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGTGCCCCTTCCTCAATTCTGTGTC[C/T]GCACTGCTCTCCAGGTGGGGCTGCG
Phenotype
MIM: 191350
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C2CD2L PubMed Links

Gene Details

Gene
C2CD2L
Gene Name
C2CD2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290474.1 146 Intron NP_001277403.1
NM_014807.4 146 Intron NP_055622.3
XM_006718948.3 146 Intron XP_006719011.1
XM_011543076.2 146 UTR 5 XP_011541378.1
XM_017018625.1 146 Intron XP_016874114.1
XM_017018626.1 146 Intron XP_016874115.1
XM_017018627.1 146 Intron XP_016874116.1
XM_017018628.1 146 UTR 5 XP_016874117.1
Gene
DPAGT1
Gene Name
dolichyl-phosphate N-acetylglucosaminephosphotransferase 1
There are no transcripts associated with this gene.

View Full Product Details