Product Details

SNP ID
rs1571858
Assay Type
Validated
NCBI dbSNP Submissions
53
Location
Chr.1:109737292 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTCCATTTTTTTGTAGACAGTGCCT[C/T]CAAGGTATAGAAGTTAGGCTCTTCC
Phenotype
MIM: 138390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSTM3 PubMed Links

Gene Details

Gene
GSTM3
Gene Name
glutathione S-transferase mu 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000849.4 Intron NP_000840.2

View Full Product Details