Product Details

SNP ID
rs13280937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123774852 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTTGTAAGGTAGTAGAATACAA[G/T]TCAATAAAGCTCTGTTTTGTACCTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM91A1 PubMed Links

Gene Details

Gene
FAM91A1
Gene Name
family with sequence similarity 91 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317917.1 Intron NP_001304846.1
NM_001317918.1 Intron NP_001304847.1
NM_144963.3 Intron NP_659400.2

View Full Product Details