Product Details
- SNP ID
-
rs4806414
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:37147886 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCTTTATGTCTGAGCAGCCCTCCA[C/T]AGTATTGGTGTACACAATTTGTTTA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF585A
PubMed Links
Gene Details
- Gene
- ZNF585A
- Gene Name
- zinc finger protein 585A
View Full Product Details