Product Details

SNP ID
rs8043748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:11752375 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTTTGAGAATCACAGCTGTGAAC[A/G]CAAAGGTCATTTGGGCAAATTGGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TXNDC11 PubMed Links

Gene Details

Gene
TXNDC11
Gene Name
thioredoxin domain containing 11
There are no transcripts associated with this gene.

Gene
ZC3H7A
Gene Name
zinc finger CCCH-type containing 7A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014153.3 Intron NP_054872.2
XM_006720877.2 Intron XP_006720940.1
XM_011522463.1 Intron XP_011520765.1
XM_011522464.2 Intron XP_011520766.1
XM_017023174.1 Intron XP_016878663.1
XM_017023175.1 Intron XP_016878664.1

View Full Product Details