Product Details

SNP ID
rs4964212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:107323653 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTACTTCACTGTGCCTTTGTATG[G/T]GCTAGTCATTGTACCTTAAACTCCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BTBD11 PubMed Links

Gene Details

Gene
BTBD11
Gene Name
BTB domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017523.1 Intron NP_001017523.1
NM_001018072.1 Intron NP_001018082.1
XM_005268645.3 Intron XP_005268702.1
XM_011537909.2 Intron XP_011536211.1
XM_011537910.2 Intron XP_011536212.1
XM_017018807.1 Intron XP_016874296.1

View Full Product Details