Product Details

SNP ID
rs1643664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:80635952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCTTCTATTACAAATGGGGAA[A/C]ATAAGAGGTTCTAGAAACAGGAGCC
Phenotype
MIM: 126060
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DHFR PubMed Links

Gene Details

Gene
DHFR
Gene Name
dihydrofolate reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000791.3 Intron NP_000782.1
NM_001290354.1 Intron NP_001277283.1
NM_001290357.1 Intron NP_001277286.1

View Full Product Details