Product Details

SNP ID
rs190421962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:80626271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTATTAAGATGGAATTGTTCTTA[C/T]TAAAGAAATAGATGAAAATGGTTAA
Phenotype
MIM: 126060
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHFR PubMed Links

Gene Details

Gene
DHFR
Gene Name
dihydrofolate reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000791.3 3756 UTR 3 NP_000782.1
NM_001290354.1 3756 UTR 3 NP_001277283.1
NM_001290357.1 3756 UTR 3 NP_001277286.1
Gene
LINC01337
Gene Name
long intergenic non-protein coding RNA 1337
There are no transcripts associated with this gene.

View Full Product Details