Product Details

SNP ID
rs751259
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:19536968 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGTGCTTATTTCTCAAAAGATGC[A/G]TCCATTTTATCCTGGAGAAAGCTTG
Phenotype
MIM: 609832
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC47A1 PubMed Links

Gene Details

Gene
SLC47A1
Gene Name
solute carrier family 47 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018242.2 Intron NP_060712.2

View Full Product Details