Product Details

SNP ID
rs832410
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:48895503 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGCTCACTTAAACTTACCAGGTTGG[C/T]CCACTCTGCTTGTCCATCCAAATCC
Phenotype
MIM: 603192
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP5G1 PubMed Links

Gene Details

Gene
ATP5G1
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C1 (subunit 9)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002027.1 Intron NP_001002027.1
NM_005175.2 Intron NP_005166.1
Gene
LOC105371814
Gene Name
uncharacterized LOC105371814
There are no transcripts associated with this gene.

View Full Product Details