Product Details
- SNP ID
-
rs1298577
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:107153172 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CTTCAATCTTTTTTCAGTTACTTAC[A/G]TCTCCACATGCTCCTCATCTGCATC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NUP62CL
PubMed Links
Gene Details
- Gene
- NUP62CL
- Gene Name
- nucleoporin 62 C-terminal like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017681.2 |
789 |
Missense Mutation |
|
|
NP_060151.2 |
View Full Product Details