Product Details

SNP ID
rs707710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75588194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCGTTCCGACCGCGGGCAGGCGG[A/C]CAGGGCGAAACCGGCGGCCGAGCCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MYO15B PubMed Links

Gene Details

Gene
MYO15B
Gene Name
myosin XVB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001309242.1 650 Missense Mutation GAC,GCC D46A NP_001296171.1
XM_017025120.1 650 Missense Mutation GAC,GCC D46A XP_016880609.1
XM_017025121.1 650 Missense Mutation GAC,GCC D46A XP_016880610.1
XM_017025122.1 650 Missense Mutation GAC,GCC D46A XP_016880611.1
XM_017025123.1 650 Missense Mutation GAC,GCC D46A XP_016880612.1
XM_017025124.1 650 Missense Mutation GAC,GCC D46A XP_016880613.1
XM_017025125.1 650 Missense Mutation GAC,GCC D46A XP_016880614.1
XM_017025126.1 650 Missense Mutation GAC,GCC D46A XP_016880615.1
XM_017025127.1 650 Missense Mutation GAC,GCC D46A XP_016880616.1
XM_017025128.1 650 Missense Mutation ACA,CCA T16P XP_016880617.1
XM_017025129.1 650 Intron XP_016880618.1
XM_017025130.1 650 Intron XP_016880619.1
XM_017025131.1 650 Intron XP_016880620.1
XM_017025132.1 650 Intron XP_016880621.1
XM_017025133.1 650 Intron XP_016880622.1
XM_017025134.1 650 Intron XP_016880623.1
XM_017025135.1 650 Intron XP_016880624.1
XM_017025136.1 650 Intron XP_016880625.1
XM_017025137.1 650 Intron XP_016880626.1
XM_017025138.1 650 Intron XP_016880627.1
XM_017025139.1 650 Intron XP_016880628.1
XM_017025140.1 650 Intron XP_016880629.1
XM_017025141.1 650 Missense Mutation GAC,GCC D46A XP_016880630.1
XM_017025142.1 650 Missense Mutation GAC,GCC D46A XP_016880631.1
XM_017025143.1 650 Intron XP_016880632.1
XM_017025144.1 650 UTR 5 XP_016880633.1

View Full Product Details