Product Details

SNP ID
rs1064060
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48936394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAGCGGGAGTCCTCAGGAAGCA[A/G]AAAGCTGGAATCAGGGAAAGCATAA
Phenotype
MIM: 103190
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ARF3 PubMed Links

Gene Details

Gene
ARF3
Gene Name
ADP ribosylation factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001659.2 3666 UTR 3 NP_001650.1
XM_005268856.1 3666 UTR 3 XP_005268913.1
XM_006719391.3 3666 Intron XP_006719454.1

View Full Product Details