Product Details

SNP ID
rs1054400
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:30782936 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GAAAAAAATAATTCACATTAAAATA[A/G]AATCTGTAGGGTGGCATAGAAGACA
Phenotype
MIM: 614853
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRLF3 PubMed Links

Gene Details

Gene
CRLF3
Gene Name
cytokine receptor like factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015986.3 2863 UTR 3 NP_057070.3
XM_011524879.1 2863 UTR 3 XP_011523181.1

View Full Product Details