Product Details

SNP ID
rs3845625
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:159296086 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATAGTGGAATATCTGTTTACTTT[C/T]TGATGAATTTTATAATGTTTATGTC
Phenotype
MIM: 147140
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FCER1A PubMed Links

Gene Details

Gene
FCER1A
Gene Name
Fc fragment of IgE receptor Ia
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002001.3 Intron NP_001992.1

View Full Product Details