Product Details

SNP ID
rs1785560
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.18:9116082 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGCAGTTTGAATTCTGTGAAACTTT[C/T]GTTCATGATCTTTACTCATTTTTTA
Phenotype
MIM: 600532
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDUFV2 PubMed Links

Gene Details

Gene
NDUFV2
Gene Name
NADH:ubiquinone oxidoreductase core subunit V2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021074.4 Intron NP_066552.2
XM_017025782.1 Intron XP_016881271.1
Gene
NDUFV2-AS1
Gene Name
NDUFV2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details