Product Details

SNP ID
rs1780706
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:37110735 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAGCATTTTGCAGGTACTCCTCTCC[C/T]GGCACGCACTTTTTTACATCGAGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MROH8 PubMed Links

Gene Details

Gene
MROH8
Gene Name
maestro heat like repeat family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152503.5 Intron NP_689716.4
NM_213631.2 Intron NP_998796.1
NM_213632.2 Intron NP_998797.2

View Full Product Details